accessibility menu, dialog, popup

UserName

CAS RN 898289-52-6

ONFFFOOFFFFF

CAS RN 898289-52-6

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 2-(morpholin-4-yl)-5-(trifluoromethyl)benzoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 2-(morpholin-4-yl)-5-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 441.28
Molecular Formula: C18H11F8NO3
InChi Key: NRNRYELFJREPET-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=C(C=CC(=C2)C(F)(F)F)N2CCOCC2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)