accessibility menu, dialog, popup

UserName

CAS RN 898289-51-5

ClNN

CAS RN 898289-51-5

IUPAC Name: 2-[3-(chloromethyl)phenyl]pyrimidine
Synonyms: 2-[3-(chloromethyl)phenyl]pyrimidine
Molecular Weight (g/mol): 204.66
Molecular Formula: C11H9ClN2
InChi Key: AUWJTKGKFPDHDU-UHFFFAOYSA-N
SMILES: ClCC1=CC=CC(=C1)C1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)