accessibility menu, dialog, popup

UserName

CAS RN 898289-37-7

OON

CAS RN 898289-37-7

IUPAC Name: 2-(oxan-4-yloxy)benzonitrile
Synonyms: 2-(oxan-4-yloxy)benzonitrile
Molecular Weight (g/mol): 203.24
Molecular Formula: C12H13NO2
InChi Key: SXNKDLAJDADKEE-UHFFFAOYSA-N
SMILES: N#CC1=CC=CC=C1OC1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)