accessibility menu, dialog, popup

UserName

CAS RN 896160-69-3

NBrH2NNH2

CAS RN 896160-69-3

IUPAC Name: 3-bromopyridine-2,5-diamine
Synonyms: 3-bromopyridine-2,5-diamine
Molecular Weight (g/mol): 188.03
Molecular Formula: C5H6BrN3
InChi Key: YJENCMIONVUOCW-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=C(N)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results