accessibility menu, dialog, popup

UserName

CAS RN 89489-53-2

IUPAC Name: thiane-4-carboxylic acid
Synonyms: thiane-4-carboxylic acid
Molecular Weight (g/mol): 146.2
Molecular Formula: C6H10O2S
InChi Key: SCKSQOIXSGPVJS-UHFFFAOYSA-N
SMILES: OC(=O)C1CCSCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results