accessibility menu, dialog, popup

UserName

BNNSH3CFF

CAS RN 892505-41-8

IUPAC Name: 2,2-difluoro-8-(methylsulfanyl)-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide
Synonyms: 2,2-difluoro-8-(methylsulfanyl)-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide
Molecular Weight (g/mol): 238.06
Molecular Formula: C10H9BF2N2S
InChi Key: OSUYHKLOJHHXAI-UHFFFAOYSA-N
SMILES: CSC1=C2C=CC=[N+]2[B-](F)(F)N2C=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results