accessibility menu, dialog, popup

UserName

CAS RN 892502-13-5

IUPAC Name: 4-[2-bromo-4-(trifluoromethyl)phenyl]morpholine
Synonyms: 4-[2-bromo-4-(trifluoromethyl)phenyl]morpholine
Molecular Weight (g/mol): 310.11
Molecular Formula: C11H11BrF3NO
InChi Key: AGOXTIRRUBMZKH-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Br)=C(C=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)