accessibility menu, dialog, popup

UserName

CAS RN 890-98-2

IUPAC Name: benzyl 2-hydroxy-2-phenylacetate
Synonyms: benzyl 2-hydroxy-2-phenylacetate
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: JFKWZVQEMSKSBU-UHFFFAOYNA-N
SMILES: OC(C(=O)OCC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results