accessibility menu, dialog, popup

UserName

H3CCH3ClCH3HO

CAS RN 89-68-9

IUPAC Name: 4-chloro-5-methyl-2-(propan-2-yl)phenol
Synonyms: chlorothymol epa pesticide chemical code 080403 caswell no. 216 C10H13ClO 氯麝酚 caniprevent [veterinary] pioral
Molecular Weight (g/mol): 184.66
Molecular Formula: C10H13ClO
InChi Key: KFZXVMNBUMVKLN-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(Cl)=C(C)C=C1O
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)