CAS RN 889939-48-4
IUPAC Name:
5-chloro-2-methoxy-4-methylbenzene-1-sulfonyl chloride
Synonyms:
5-chloro-2-methoxy-4-methylbenzenesulfonyl chloride
Molecular Weight (g/mol):
255.11
Molecular Formula:
C8H8Cl2O3S
InChi Key:
BJRJHJZVSWZXIP-UHFFFAOYSA-N
SMILES:
COC1=C(C=C(Cl)C(C)=C1)S(Cl)(=O)=O
1
–
1
of
1
results
1
Promotions Available
5-Chloro-2-methoxy-4-methylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
5-Chloro-2-methoxy-4-methylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
CAS: 889939-48-4 Molecular Formula: C8H8Cl2O3S Molecular Weight (g/mol): 255.109 MDL Number: MFCD08445709 InChI Key: BJRJHJZVSWZXIP-UHFFFAOYSA-N Synonym: 5-chloro-2-methoxy-4-methylbenzenesulfonylchloride,5-chloro-2-methoxy-4-methylbenzene-1-sulfonyl chloride,pubchem5095,acmc-20am4m,5-chloro-2-methoxy-4-methylphenylsulfonyl chloride,chloro 5-chloro-2-methoxy-4-methylphenyl sulfone,5-chloranyl-2-methoxy-4-methyl-benzenesulfonyl chloride PubChem CID: 17389862 IUPAC Name: 5-chloro-2-methoxy-4-methylbenzenesulfonyl chloride SMILES: CC1=CC(=C(C=C1Cl)S(=O)(=O)Cl)OC