accessibility menu, dialog, popup

UserName

CAS RN 885698-94-2

BOOCH3H3CCH3CH3NNH3C

CAS RN 885698-94-2

IUPAC Name: 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole
Synonyms: 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole
Molecular Weight (g/mol): 258.13
Molecular Formula: C14H19BN2O2
InChi Key: OZHFBNULPUBRJF-UHFFFAOYSA-N
SMILES: CN1N=CC2=C(C=CC=C12)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)