accessibility menu, dialog, popup

UserName

NH2NI

CAS RN 88511-27-7

IUPAC Name: 3-iodopyridin-4-amine
Synonyms: 3-iodopyridin-4-amine
Molecular Weight (g/mol): 220.01
Molecular Formula: C5H5IN2
InChi Key: ZGOCEDVVZKFHSY-UHFFFAOYSA-N
SMILES: NC1=CC=NC=C1I
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)