accessibility menu, dialog, popup

UserName

CH3NONHNH3C

CAS RN 884507-32-8

IUPAC Name: methyl({[4-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methyl})amine
Synonyms: methyl({[4-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methyl})amine
Molecular Weight (g/mol): 203.25
Molecular Formula: C11H13N3O
InChi Key: MUCOLQGVWQXQMX-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(C=C1)C1=NC(C)=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)