accessibility menu, dialog, popup

UserName

Uncover New Spotlight Deals and Get Up to 50% Off

Save Today

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 884507-29-3

SNNOOH

CAS RN 884507-29-3

IUPAC Name: 2-(thiomorpholin-4-yl)pyridine-4-carboxylic acid
Synonyms: 2-(thiomorpholin-4-yl)pyridine-4-carboxylic acid
Molecular Weight (g/mol): 224.28
Molecular Formula: C10H12N2O2S
InChi Key: DDAWRDLRTZFVHD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=NC=C1)N1CCSCC1

Narrow Results

Narrow Results

  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.