CAS RN 884494-44-4
IUPAC Name:
(2-chloro-4-iodopyridin-3-yl)methanol
Synonyms:
(2-chloro-4-iodopyridin-3-yl)methanol
Molecular Weight (g/mol):
269.47
Molecular Formula:
C6H5ClINO
InChi Key:
BZQRQPAKTCMYIA-UHFFFAOYSA-N
SMILES:
OCC1=C(I)C=CN=C1Cl
1
–
1
of
1
results
1
Promotions Available
2-Chloro-4-iodo-3-pyridinemethanol, 97%, Thermo Scientific Chemicals
2-Chloro-4-iodo-3-pyridinemethanol, 97%, Thermo Scientific Chemicals
CAS: 884494-44-4 Molecular Formula: C6H5ClINO Molecular Weight (g/mol): 269.466 MDL Number: MFCD03095296 InChI Key: BZQRQPAKTCMYIA-UHFFFAOYSA-N Synonym: 2-chloro-4-iodopyridin-3-yl methanol,2-chloro-4-iodo-3-pyridinemethanol,2-chloro-3-hydroxymethyl-4-iodopyridine,2-chloro-4-iodo-pyridin-3-yl-methanol,acmc-20anzo,pubchem17089,3-pyridinemethanol,2-chloro-4-iodo,2-chloro-4-iodo-3-pyridyl methan-1-ol PubChem CID: 40418628 IUPAC Name: (2-chloro-4-iodopyridin-3-yl)methanol SMILES: C1=CN=C(C(=C1I)CO)Cl