accessibility menu, dialog, popup

UserName

CAS RN 876316-33-5

IUPAC Name: 4-{[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}morpholine
Synonyms: 4-{[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}morpholine
Molecular Weight (g/mol): 303.21
Molecular Formula: C17H26BNO3
InChi Key: WCNXVDLZTGODQW-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=CC=C1CN1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)