accessibility menu, dialog, popup

UserName

CH3H3CSiH3CH3CCH3CH3NBr

CAS RN 87630-36-2

IUPAC Name: 3-bromo-1-[tris(propan-2-yl)silyl]-1H-pyrrole
Synonyms: 3-bromo-1-(triisopropylsilyl)pyrrole
Molecular Weight (g/mol): 302.33
Molecular Formula: C13H24BrNSi
InChi Key: CNAUTMPRKBSDLH-UHFFFAOYSA-N
SMILES: CC(C)[Si](C(C)C)(C(C)C)N1C=CC(Br)=C1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results