accessibility menu, dialog, popup

UserName

CAS RN 873537-32-7

IUPAC Name: tert-butyl N-(4-cyanocyclohexyl)carbamate
Synonyms: tert-butyl N-(4-cyanocyclohexyl)carbamate
Molecular Weight (g/mol): 224.3
Molecular Formula: C12H20N2O2
InChi Key: QHZJRHRMFIDRFL-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CCC(CC1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)