accessibility menu, dialog, popup

UserName

CH3SOOOON

CAS RN 87331-48-4

IUPAC Name: 5-methanesulfonyl-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Synonyms: 5-methanesulfonyl-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Molecular Weight (g/mol): 319.38
Molecular Formula: C16H17NO4S
InChi Key: DPJWIXMOCGJKDG-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC2=CC3=C4N(CCC3)CCCC4=C2OC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results