accessibility menu, dialog, popup

UserName

CAS RN 872578-72-8

IUPAC Name: N-(2-{[5-acetamido-4-hydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy}-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl)acetamide
Synonyms: N-(2-{[5-acetamido-4-hydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy}-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl)acetamide
Molecular Weight (g/mol): 545.5
Molecular Formula: C22H31N3O13
InChi Key: HWBFEVWOQMUQIE-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(O)C(O)C(CO)OC1OC1C(O)C(NC(C)=O)C(OC2=CC=C(C=C2)[N+]([O-])=O)OC1CO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results