accessibility menu, dialog, popup

UserName

CAS RN 87254-55-5

H3CONCOH3C

CAS RN 87254-55-5

IUPAC Name: 7-isocyanato-2,2-dimethyl-2,3-dihydro-1-benzofuran
Synonyms: 7-isocyanato-2,2-dimethyl-3H-1-benzofuran
Molecular Weight (g/mol): 189.21
Molecular Formula: C11H11NO2
InChi Key: JTJCJMYVKXVWIJ-UHFFFAOYSA-N
SMILES: CC1(C)CC2=C(O1)C(=CC=C2)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)