CAS RN 87219-80-5
IUPAC Name:
(3,3-diethoxyprop-1-yn-1-yl)trimethylsilane
Synonyms:
(3,3-diethoxyprop-1-yn-1-yl)trimethylsilane
Molecular Weight (g/mol):
200.35
Molecular Formula:
C10H20O2Si
InChi Key:
MDRSJACTZKYOAL-UHFFFAOYSA-N
SMILES:
CCOC(OCC)C#C[Si](C)(C)C
1
–
1
of
1
results
1
Promotions Available
3-(Trimethylsilyl)propiolaldehyde diethyl acetal, 97%, Thermo Scientific Chemicals
3-(Trimethylsilyl)propiolaldehyde diethyl acetal, 97%, Thermo Scientific Chemicals
CAS: 87219-80-5 Molecular Formula: C10H20O2Si Molecular Weight (g/mol): 200.353 MDL Number: MFCD08275372 InChI Key: MDRSJACTZKYOAL-UHFFFAOYSA-N Synonym: 3,3-diethoxyprop-1-yn-1-yl trimethylsilane,1-trimethylsilyl-3,3-diethoxy-1-propyne,3-trimethylsilyl propiolaldehyde diethyl acetal,3-trimethylsilylpropargyl aldehyde diethyl acetal PubChem CID: 11206442 IUPAC Name: 3,3-diethoxyprop-1-ynyl(trimethyl)silane SMILES: CCOC(C#C[Si](C)(C)C)OCC