CAS RN 87184-99-4
IUPAC Name:
4-[(tert-butyldimethylsilyl)oxy]butan-1-ol
Synonyms:
4-[(tert-butyldimethylsilyl)oxy]butan-1-ol
Molecular Weight (g/mol):
204.39
Molecular Formula:
C10H24O2Si
InChi Key:
IJEMXJANZPVITP-UHFFFAOYSA-N
SMILES:
CC(C)(C)[Si](C)(C)OCCCCO
1
–
1
of
1
results
1
Promotions Available
4-tert-Butyldimethylsiloxy-1-butanol, 97%
4-tert-Butyldimethylsiloxy-1-butanol, 97%
CAS: 87184-99-4 Molecular Formula: C10H24O2Si Molecular Weight (g/mol): 204.39 MDL Number: MFCD04974070 InChI Key: IJEMXJANZPVITP-UHFFFAOYSA-N Synonym: 4-tert-butyldimethylsilyl oxy butan-1-ol,4-dimethyl-tert-butylsilyl oxybutan-1-ol,4-dimethyl-tert-butylsilyloxy butan-1-ol,4-tert-butyldimethylsilyl oxy-1-butanol,1-butanol, 4-1,1-dimethylethyl dimethylsilyl oxy,acmc-20amup,4-t-butyldimethylsiloxy-1-butanol,4-t-butyldimethylsilyloxy-1-butanol,4-tert-butyldimethylsiloxy-1-butanol PubChem CID: 186403 IUPAC Name: 4-[tert-butyl(dimethyl)silyl]oxybutan-1-ol SMILES: CC(C)(C)[Si](C)(C)OCCCCO