CAS RN 87-17-2
CAS RN 87-17-2
IUPAC Name:
2-hydroxy-N-phenylbenzamide
Synonyms:
salicylanilide salinide salicylic acid anilide 2-N-phenylcarboxamidophenol
Molecular Weight (g/mol):
213.24
Molecular Formula:
C13H11NO2
InChi Key:
WKEDVNSFRWHDNR-UHFFFAOYSA-N
SMILES:
OC1=CC=CC=C1C(=O)NC1=CC=CC=C1
1
–
1
of
1
results
1
Salicylanilide 98.0+%, TCI America™
CAS: 87-17-2 Molecular Formula: C13H11NO2 Molecular Weight (g/mol): 213.24 MDL Number: MFCD00002212 InChI Key: WKEDVNSFRWHDNR-UHFFFAOYSA-N Synonym: salicylanilide,n-phenylsalicylamide,salinide,salicylanilid,salifebrin,salinidol,ansadol,salicylic acid anilide,salnide,benzamide, 2-hydroxy-n-phenyl PubChem CID: 6872 ChEBI: CHEBI:239133 IUPAC Name: 2-hydroxy-N-phenylbenzamide SMILES: OC1=CC=CC=C1C(=O)NC1=CC=CC=C1