accessibility menu, dialog, popup

UserName

H3COOOCH3

CAS RN 87-05-8

IUPAC Name: 7-ethoxy-4-methyl-2H-chromen-2-one
Synonyms: 7-ethoxy-4-methylchromen-2-one
Molecular Weight (g/mol): 204.23
Molecular Formula: C12H12O3
InChi Key: NKRISXMDKXBVRJ-UHFFFAOYSA-N
SMILES: CCOC1=CC=C2C(C)=CC(=O)OC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results