CAS RN 86873-60-1
IUPAC Name:
5-chloropyridine-2-carboxylic acid
Synonyms:
5-chloropyridine-2-carboxylic acid
Molecular Weight (g/mol):
157.55
Molecular Formula:
C6H4ClNO2
InChi Key:
GJLOKYIYZIOIPN-UHFFFAOYSA-N
SMILES:
OC(=O)C1=NC=C(Cl)C=C1
1
–
1
of
1
results
1
Promotions Available
5-Chloropyridine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
5-Chloropyridine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
CAS: 86873-60-1 Molecular Formula: C6H4ClNO2 Molecular Weight (g/mol): 157.553 MDL Number: MFCD04114192 InChI Key: GJLOKYIYZIOIPN-UHFFFAOYSA-N Synonym: 5-chloropicolinic acid,5-chloro-2-picolinic acid,5-chloro-2-pyridinecarboxylic acid,5-chloro-pyridine-2-carboxylic acid,5-chloro-2-carboxypyridine,2-carboxy-5-chloropyridine,5-chloropyridine-2-carboxylicacid,2-pyridinecarboxylic acid, 5-chloro,pubchem10379,5-chloro-picolinic acid PubChem CID: 2762872 IUPAC Name: 5-chloropyridine-2-carboxylic acid SMILES: C1=CC(=NC=C1Cl)C(=O)O