accessibility menu, dialog, popup

UserName

CAS RN 86770-68-5

IUPAC Name: 14-azido-3,6,9,12-tetraoxatetradecan-1-ol
Synonyms: 14-azido-3,6,9,12-tetraoxatetradecan-1-ol
Molecular Weight (g/mol): 263.29
Molecular Formula: C10H21N3O5
InChi Key: JTGGTGKXQQGEHB-UHFFFAOYSA-N
SMILES: OCCOCCOCCOCCOCCN=[N+]=[N-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results