accessibility menu, dialog, popup

UserName

NNBr

CAS RN 864377-28-6

IUPAC Name: 4-(3-bromophenyl)-2,6-diphenylpyrimidine
Synonyms: 4-(3-bromophenyl)-2,6-diphenylpyrimidine
Molecular Weight (g/mol): 387.28
Molecular Formula: C22H15BrN2
InChi Key: BPMSGKUGXMWVBH-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)C1=NC(=NC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results