accessibility menu, dialog, popup

UserName

HOON

CAS RN 86-59-9

IUPAC Name: quinoline-8-carboxylic acid
Synonyms: 8-quinolinecarboxylic acid
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: QRDZFPUVLYEQTA-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2N=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)