accessibility menu, dialog, popup

UserName

CAS RN 858-46-8

IUPAC Name: tris(1,1,2,2,2-pentafluoroethyl)-1,3,5-triazine
Synonyms: tris(1,1,2,2,2-pentafluoroethyl)-1,3,5-triazine
Molecular Weight (g/mol): 435.1
Molecular Formula: C9F15N3
InChi Key: MQBPAXAOMVELQG-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C1=NC(=NC(=N1)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results