accessibility menu, dialog, popup

UserName

CAS RN 85553-53-3

IUPAC Name: 3-(pyridin-2-yl)benzaldehyde
Synonyms: 3-(pyridin-2-yl)benzaldehyde
Molecular Weight (g/mol): 183.21
Molecular Formula: C12H9NO
InChi Key: SAPNGHSAYQXRPG-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC(=C1)C1=CC=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results