CAS RN 852180-63-3
IUPAC Name:
[5-(furan-2-yl)-1,2-oxazol-3-yl]methanol
Synonyms:
[5-(furan-2-yl)-1,2-oxazol-3-yl]methanol
Molecular Weight (g/mol):
165.15
Molecular Formula:
C8H7NO3
InChi Key:
NBBDGNIUCJFJPO-UHFFFAOYSA-N
SMILES:
OCC1=NOC(=C1)C1=CC=CO1
1
–
1
of
1
results
1
[5-(2-Furyl)isoxazol-3-yl]methanol, ≥97%, Thermo Scientific™
CAS: 852180-63-3 Molecular Formula: C8H7NO3 Molecular Weight (g/mol): 165.148 MDL Number: MFCD07772820 InChI Key: NBBDGNIUCJFJPO-UHFFFAOYSA-N Synonym: 5-2-furyl isoxazol-3-yl methanol,5-furan-2-yl-1,2-oxazol-3-yl methanol,5-furan-2-yl isoxazol-3-yl methanol,5-2-furyl isoxazol-3-yl methan-1-ol,5-2-furanyl-3-isoxazolylmethanol,5-2-furyl-3-isoxazolyl methanol,5-furan-2-yl-isoxazol-3-yl-methanol PubChem CID: 7164591 IUPAC Name: [5-(furan-2-yl)-1,2-oxazol-3-yl]methanol SMILES: C1=COC(=C1)C2=CC(=NO2)CO