accessibility menu, dialog, popup

UserName

CAS RN 85116-37-6

BCH3CH3CH3CH3H3CH3CCl

CAS RN 85116-37-6

IUPAC Name: chlorobis({2,6,6-trimethylbicyclo[3.1.1]heptan-3-yl})borane
Synonyms: chlorobis({2,6,6-trimethylbicyclo[3.1.1]heptan-3-yl})borane
Molecular Weight (g/mol): 320.75
Molecular Formula: C20H34BCl
InChi Key: PSEHHVRCDVOTID-UHFFFAOYNA-N
SMILES: CC1C2CC(CC1B(Cl)C1CC3CC(C1C)C3(C)C)C2(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results