accessibility menu, dialog, popup

UserName

CAS RN 850567-92-9

IUPAC Name: (4-methylcyclohex-1-en-1-yl)boronic acid
Synonyms: 4-methylcyclohex-1-en-1-ylboronic acid
Molecular Weight (g/mol): 139.99
Molecular Formula: C7H13BO2
InChi Key: SJZMIZIVYIOMIW-UHFFFAOYNA-N
SMILES: CC1CCC(=CC1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results