accessibility menu, dialog, popup

UserName

CH3HNONBr

CAS RN 845305-87-5

IUPAC Name: 5-bromo-N-methylpyridine-2-carboxamide
Synonyms: 5-bromo-N-methylpyridine-2-carboxamide
Molecular Weight (g/mol): 215.05
Molecular Formula: C7H7BrN2O
InChi Key: OHYCXPXWHPWYFX-UHFFFAOYSA-N
SMILES: CNC(=O)C1=NC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results