accessibility menu, dialog, popup

UserName

CAS RN 84366-81-4

IUPAC Name: disodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[(2R,3S,4S)-5-{7,8-dimethyl-2,4-dioxo-2H,3H,4H,10H-benzo[g]pteridin-10-yl}-2,3,4-trihydroxypentyl phosphonato]oxy}phosphonate
Synonyms: disodium [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl [(2R,3S,4S)-5-{7,8-dimethyl-2,4-dioxo-3H-benzo[g]pteridin-10-yl}-2,3,4-trihydroxypentyl phosphonato]oxyphosphonate
Molecular Weight (g/mol): 829.52
Molecular Formula: C27H31N9Na2O15P2
InChi Key: XLRHXNIVIZZOON-SJFBGTSINA-L
SMILES: [Na+].[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)N3C=NC4=C(N)N=CN=C34)C3=NC(=O)NC(=O)C3=NC2=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results