accessibility menu, dialog, popup

UserName

CAS RN 84365-55-9

HCClTiClCl

CAS RN 84365-55-9

IUPAC Name: 1H-inden-1-ide; trichlorotitaniumylium
Synonyms: 1H-inden-1-ide; trichlorotitaniumylium
Molecular Weight (g/mol): 269.37
Molecular Formula: C9H7Cl3Ti
InChi Key: MDTDQDVMQBTXST-UHFFFAOYSA-K
SMILES: Cl[Ti+](Cl)Cl.[CH-]1C=CC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results