accessibility menu, dialog, popup

UserName

CAS RN 84163-42-8

NOHOO

CAS RN 84163-42-8

IUPAC Name: 1-formylpiperidine-4-carboxylic acid
Synonyms: 1-formylpiperidine-4-carboxylic acid
Molecular Weight (g/mol): 157.17
Molecular Formula: C7H11NO3
InChi Key: IZNTWTMLHCGAJU-UHFFFAOYSA-N
SMILES: OC(=O)C1CCN(CC1)C=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results