accessibility menu, dialog, popup

UserName

CAS RN 83972-01-4

IUPAC Name: magnesium(2+) bis(3-[(4-nitrophenyl)methoxy]-3-oxopropanoate)
Synonyms: magnesium(2+) bis(3-[(4-nitrophenyl)methoxy]-3-oxopropanoate)
Molecular Weight (g/mol): 500.66
Molecular Formula: C20H16MgN2O12
InChi Key: WAFDWKYNSTVECG-UHFFFAOYSA-L
SMILES: [Mg++].[O-]C(=O)CC(=O)OCC1=CC=C(C=C1)[N+]([O-])=O.[O-]C(=O)CC(=O)OCC1=CC=C(C=C1)[N+]([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results