CAS RN 83140-94-7
IUPAC Name:
[3-(1H-pyrrol-1-yl)phenyl]methanol
Synonyms:
[3-(pyrrol-1-yl)phenyl]methanol
Molecular Weight (g/mol):
173.22
Molecular Formula:
C11H11NO
InChi Key:
QQXDXYAGEXWXQU-UHFFFAOYSA-N
SMILES:
OCC1=CC(=CC=C1)N1C=CC=C1
1
–
3
of
3
results
1
[3-(1H-Pyrrol-1-yl)phenyl]methanol, 97%, Thermo Scientific™
CAS: 83140-94-7 Molecular Formula: C11H11NO Molecular Weight (g/mol): 173.215 MDL Number: MFCD03086142 InChI Key: QQXDXYAGEXWXQU-UHFFFAOYSA-N Synonym: 3-1h-pyrrol-1-yl phenyl methanol,3-pyrrol-1-yl phenyl methanol,3-1-pyrrolyl benzyl alcohol,3-pyrrol-1-ylphenyl methanol,3-pyrrolylphenyl methan-1-ol,3-pyrrol-1-yl phenylmethanol,3-1h-pyrrol-1-yl phenylmethanol,benzenemethanol, 3-1h-pyrrol-1-yl PubChem CID: 2776531 IUPAC Name: (3-pyrrol-1-ylphenyl)methanol SMILES: C1=CN(C=C1)C2=CC=CC(=C2)CO