accessibility menu, dialog, popup

UserName

H3CNHHN

CAS RN 83030-08-4

IUPAC Name: N-methylpyrrolidin-3-amine
Synonyms: N-methylpyrrolidin-3-amine
Molecular Weight (g/mol): 100.17
Molecular Formula: C5H12N2
InChi Key: NGZYRKGJWYJGRS-UHFFFAOYNA-N
SMILES: CNC1CCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results