CAS RN 824-40-8
IUPAC Name:
2-carboxypyridin-1-ium-1-olate
Synonyms:
2-carboxypyridin-1-ium-1-olate
Molecular Weight (g/mol):
139.11
Molecular Formula:
C6H5NO3
InChi Key:
FHYMLBVGNFVFBT-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=CC=[N+]1[O-]
1
–
1
of
1
results
1
Promotions Available
Picolinic acid N-oxide, 97%, Thermo Scientific™
Picolinic acid N-oxide, 97%, Thermo Scientific™
CAS: 824-40-8 Molecular Formula: C6H5NO3 Molecular Weight (g/mol): 139.11 MDL Number: MFCD00006198 InChI Key: FHYMLBVGNFVFBT-UHFFFAOYSA-N Synonym: picolinic acid n-oxide,2-carboxypyridin-1-ium-1-olate,pyridine-2-carboxylic acid 1-oxide,picolinic acid 1-oxide,2-carboxypyridine n-oxide,2-pyridinecarboxylic acid, 1-oxide,chembl73173,2-pyridinecarboxylicacid, 1-oxide,picolinate n-oxide,1-oxopicolinic acid PubChem CID: 69986 IUPAC Name: 1-oxidopyridin-1-ium-2-carboxylic acid SMILES: C1=CC=[N+](C(=C1)C(=O)O)[O-]