accessibility menu, dialog, popup

UserName

NOSOOClSOOON

CAS RN 82354-19-6

IUPAC Name: 16-[4-(chlorosulfonyl)-2-sulfonatophenyl]-3-oxa-9λ⁵,23-diazaheptacyclo[17.7.1.1⁵,⁹.0²,¹⁷.0⁴,¹⁵.0²³,²⁷.0¹³,²⁸]octacosa-1,4,9(28),13,15,17,19(27)-heptaen-9-ylium
Synonyms: texas red sulforhodamine 101 acid chloride sulforhodamine 101 sulfonyl chloride
Molecular Weight (g/mol): 625.15
Molecular Formula: C31H29ClN2O6S2
InChi Key: MPLHNVLQVRSVEE-UHFFFAOYSA-N
SMILES: [O-]S(=O)(=O)C1=C(C=CC(=C1)S(Cl)(=O)=O)C1=C2C=C3CCC[N+]4=C3C(CCC4)=C2OC2=C3CCCN4CCCC(C=C12)=C34

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results