accessibility menu, dialog, popup

UserName

CAS RN 823-73-4

ONOOBr

CAS RN 823-73-4

IUPAC Name: 2-bromo-5-nitrofuran
Synonyms: 2-bromo-5-nitrofuran
Molecular Weight (g/mol): 191.97
Molecular Formula: C4H2BrNO3
InChi Key: QGTDMAKRJHGXOF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Br)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results