accessibility menu, dialog, popup

UserName

H3CCH3H3C

CAS RN 816-79-5

IUPAC Name: 3-ethylpent-2-ene
Synonyms: 3-ethyl-2-pentene
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: XMYFZAWUNVHVGI-UHFFFAOYSA-N
SMILES: CCC(CC)=CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results