accessibility menu, dialog, popup

UserName

CAS RN 81542-51-0

IUPAC Name: 4-iodo-1H-pyrazol-5-amine
Synonyms: 4-iodo-2H-pyrazol-3-amine
Molecular Weight (g/mol): 208.99
Molecular Formula: C3H4IN3
InChi Key: CPVCVVMJPIOKMN-UHFFFAOYSA-N
SMILES: NC1=C(I)C=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results