accessibility menu, dialog, popup

UserName

CH3(S)BrO(R)CH3Br

CAS RN 815-60-1

IUPAC Name: (2R,4S)-2,4-dibromopentan-3-one
Synonyms: (2R,4S)-2,4-dibromopentan-3-one
Molecular Weight (g/mol): 243.93
Molecular Formula: C5H8Br2O
InChi Key: UOPIOAUZQKSZRO-ZXZARUISSA-N
SMILES: C[C@@H](Br)C(=O)[C@H](C)Br
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results