accessibility menu, dialog, popup

UserName

CAS RN 80866-95-1

IUPAC Name: 3-[(1H-pyrrol-1-yl)methyl]pyridine
Synonyms: 3-(pyrrol-1-ylmethyl)pyridine
Molecular Weight (g/mol): 158.2
Molecular Formula: C10H10N2
InChi Key: PJVWPGGKILHMKW-UHFFFAOYSA-N
SMILES: C(N1C=CC=C1)C1=CC=CN=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results