accessibility menu, dialog, popup

UserName

HO(S)N(S)OH(R)OH(R)OH(R)

CAS RN 79831-76-8

IUPAC Name: (1S,6S,7R,8R,8aR)-octahydroindolizine-1,6,7,8-tetrol
Synonyms: castanospermine 6-epicastanospermine castinospermine 6,7-diepicastanospermine
Molecular Weight (g/mol): 189.21
Molecular Formula: C8H15NO4
InChi Key: JDVVGAQPNNXQDW-TVNFTVLESA-N
SMILES: O[C@H]1CCN2C[C@H](O)[C@@H](O)[C@H](O)[C@@H]12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results