accessibility menu, dialog, popup

UserName

H3CNFPFFFFF

CAS RN 797789-00-5

IUPAC Name: 1-hexylpyridin-1-ium; hexafluoro-λ⁵-phosphanuide
Synonyms: 1-hexylpyridin-1-ium hexafluorophosphate
Molecular Weight (g/mol): 309.24
Molecular Formula: C11H18F6NP
InChi Key: DKRTWJPQQYAIHH-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CCCCCC[N+]1=CC=CC=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results